missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SHOWA™ N-DEX™ 9700PF Nitrile Gloves

Powder free nitrile gloves offer tactile precision and high-capacity protection in a wide range of applications.

$17.72 - $17.87

Specifications

Certifications/Compliance FDA 21 CFR 170-199
Certifications/Compliance (Europe) EN 420: 2003 + A1:2009
Accelerator Free Yes
Applications Industrial, Laboratory
Color Black
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-168-935
View Documents
SHOWA™
9700PFS
Small
Pack of 50 Each for $17.72
Case of 20 Pack for $323.60
 
29-636-0116
View Documents
SHOWA™
9700PFM08
Medium
Pack of 50 Each for $17.87
Case of 20 Pack for $326.20
 
29-636-0115
View Documents
SHOWA™
9700PFL09
Large
Pack of 50 Each for $17.83
Case of 20 Pack for $325.60
 
19-168-938
View Documents
SHOWA™
9700PFXL
X-Large
Pack of 50 Each for $17.72
Case of 20 Pack for $323.60
 
Description

Description

  • Thick, 100% nitrile construction for enhanced barrier protection
  • High-capacity protection for a wide variety of applications
  • Rolled cuff prevents dirt and debris from entering the glove
  • Form-fitting design for increased sensitivity
  • Accelerator-free
  • Powder free
  • Made in the USA
Specifications

Specifications

FDA 21 CFR 170-199
Yes
Black
No Texture
Nitrile
N-DEX™ 9700
28 cm
EN 420: 2003 + A1:2009
Industrial, Laboratory
Single-use
Yes
Powder Free
11 in.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'