Siemens Healthineers Chek-Stix™ Combo Pak Urinalysis Control Strips DFS Item
Positive and negative controls
Supplier:  Siemens Healthineers 10910310483
Catalog No. 23-038104
Explore available promotions
Description
Description
Positive and negative controls
  • Convenient combination kit of positive and negative control strips for urinalysis testing
  • Chek-Stix provides positive, negative, or defined results for bilirubin, blood, glucose, ketones, leukocytes, nitrite, pH, protein, specific gravity, and urobilinogen
  • Control strips are reconstituted with distilled water to provide results; remain stable for up to eight hours
Order Info
Packaged as a set with one bottle each of 25 strips, positive and negative.
Specifications
Specifications
Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity, Urobilinogen
Urinalysis
25 Pack