Siemens Healthineers Urinalysis Reagent Test Strips
Single-use, plastic strips with multiple reagent test areas
$145.71 - $145.71
Specifications
Disposable | Single-use |
---|---|
For Use With (Application) | Urinalysis, in-vitro diagnostic use |
Catalog Number | Mfr. No. | Detectable Analytes | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Detectable Analytes | Quantity | Price | Quantity | ||||
AM2830
|
Siemens Healthineers
10910324060 |
Blood Urea, Nitrogen | 25 Pack |
Pack of 25 Each for $145.71
|
|
||||
Description
- Ready to use and entirely disposable
- Dip and read visually for results
- Some strips can be read instrumentally on Clinitek™ 50, 100, 200, 200+, 500, Advantus, and Status Family of Urine Chemistry analyzers
Specifications
Single-use | |
Urinalysis, in-vitro diagnostic use |