Epredia™ Signature Series™ Cyto-Stain™
Achieve results similar to conventional Papanicolaou stains with Epredia™ Signature Series Cyto-Stain™, a uniquely formulated cytoplasmic stain
Supplier: Epredia™ 7511
Description
- Use for all gynecological and nongynecological procedures
- Reduces staining time
- Cost-effective, as user performs only one counterstain
- Cyto-Stain provides a full range of familiar cytoplasmic colors and control over the staining intensities
- Cyto-Stain G is a modification to produce greener cyanophilic hues in intermediate and basal cells
Specifications
Cytostain | |
4 pt. |