missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Epredia™ Signature Series™ Hematoxylin 7211
Yield results that exceed those of Gill or Harris Hematoxylins with this uniquely formulated product.
Supplier: Epredia™ 7211
Description
- No filtration required
- Stains up to 2500 slides per pint
- Crisp, well-defined nuclear detail
Available in 5 liter packaging
- Reduces plastic by 90%
- Increases stain stability
- Reduces messy stain spills
- Saves 35% storage space versus traditional packaging
Specifications
Hematoxylin | |
1 pt. |