missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silica 100-200μm, 60Å, EcoChrom™, MP Biomedicals™
For Chromatography; Chemical Synthesis
$6428.75 - $6428.75
Specifications
Content And Storage | Store at Room Temperature. |
---|---|
Type | Silica |
Physical Form | Powder |
MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
Density | 2g/cm3 at 20°C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN02761
|
MP Biomedicals Inc
0202761 |
25 kg |
Each for $6,428.75
|
|
|||||
Specifications
Store at Room Temperature. | |
Powder | |
2g/cm3 at 20°C |
Silica | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
7631-86-9 |