Silica gel, C18-RP, 17%C, ca.0,8mmol/g, part. size: 40-63μ, Thermo Scientific Chemicals
Supplier:  Thermo Scientific Chemicals 360122500
 View more versions of this product
Catalog No. AC360122500
Explore available promotions
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatography
Specifications
Specifications
Silica gel, C18-RP, 17%C
250 g
Octadecylsilicate
VYTRPUAMVCWPLO-UHFFFAOYSA-N
dioxosilane;trichloro(octadecyl)silane
172932
71889-02-6
C18H37Cl3O2Si2
Solubility in water: insoluble.
CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl.O=[Si]=O
448.012
Glass Bottle
Chemical Identifiers
71889-02-6
448.012
Octadecylsilicate
dioxosilane;trichloro(octadecyl)silane
C18H37Cl3O2Si2
VYTRPUAMVCWPLO-UHFFFAOYSA-N
172932
CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl.O=[Si]=O
SDS Documents