missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silica Gel Desiccant (100-200 Mesh/923, Chromatographic Grade), Fisher Chemical™
Silica gel Grade 923 conforms to the specifications of ASTM D1319 and ASTM D2549.
Supplier: Fisher Chemical S679500
Description
- Sold as a case of 3 boxes - each box contains 24 x 10 g pouches
- Suitable for use in Fluorescent Indicator Adsorption (FIA) test
- Conforms to the specifications of ASTM D1319 and ASTM D2549
- Surface area within the range of 430 - 530 m2/g
- This grade of silica gel is also available in other sizes:
- 500 g (S679500)
- 2.5 kg (S679212)
- 25 g (S67925)
Specifications
1710°C | |
Solid | |
White | |
100-200 Mesh | |
2.1g/cm³ | |
500 g | |
O2Si | |
Inclusive between 4.5% | |
5.0% max. | |
O=[Si]=O | |
60.08 | |
CHEBI:30563 | |
Inclusive between 430m2/g |
2230°C | |
5.5 to 7.0 (for 5% slurry) | |
Silica Gel Desiccant | |
7631-86-9 | |
Silica Gel | |
Pass Test | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
Poly Bottle | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
silanedione | |
24261 | |
Chromatography |
Chemical Identifiers
7631-86-9 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
CHEBI:30563 | |
O=[Si]=O |
O2Si | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
24261 | |
silanedione |