Silica Gel Desiccant Indicating(Grade 48, 4-10 Mesh/Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical S162500
Specifications
Solid | |
Silica Gel Desiccant, Indicating | |
112926-00-8,7646-79-9 | |
500 g | |
SiO2 | |
4 to 10 Mesh | |
Pass Test |
Blue | |
4-10 Mesh, Grade 48 | |
Silica Gel | |
Pass Test | |
MFCD00011232 | |
Poly Bottle | |
Certified ACS |