Learn More
Silica gel, TLC high purity grade, with gypsum binder, Thermo Scientific Chemicals
TLC plate preparation
$89.37 - $344.34
Chemical Identifiers
CAS | 63231-67-4 |
---|---|
Molecular Formula | H2O3Si |
Molecular Weight (g/mol) | 78.098 |
MDL Number | MFCD00011232 |
InChI Key | IJKVHSBPTUYDLN-UHFFFAOYSA-N |
PubChem CID | 14768 |
IUPAC Name | dihydroxy(oxo)silane |
SMILES | O[Si](=O)O |
Description
TLC plate preparation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
63231-67-4 | |
78.098 | |
IJKVHSBPTUYDLN-UHFFFAOYSA-N | |
dihydroxy(oxo)silane |
H2O3Si | |
MFCD00011232 | |
14768 | |
O[Si](=O)O |
Specifications
12 Micron APS,S.A. 500-600m | |
63231-67-4 | |
50 g | |
MFCD00011232 | |
IJKVHSBPTUYDLN-UHFFFAOYSA-N | |
dihydroxy(oxo)silane | |
14768 | |
500 to 600m2/g |
6 to 7 | |
Silica Gel | |
H2O3Si | |
500 to 600m2/g, 60Å | |
O[Si](=O)O | |
78.098 | |
TLC High Purity | |
Silica gel, With gypsum binder |
Safety and Handling
GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 231-545-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only