missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silica Gel, With Indicator (Orange gel), MilliporeSigma™
Solution for histology
Supplier: Technidata 1019695000
Specifications
>1,000°C | |
7.3 | |
5 kg |
Orange | |
Granules | |
Plastic Bottle |
Chemical Identifiers
0.0 |