Learn More
Silicon dioxide, 99.998%, (trace metal basis), Thermo Scientific Chemicals
For ultra trace analysis
Supplier: Thermo Scientific Chemicals 437150050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisSpecifications
Silicon dioxide | |
14808-60-7 | |
Solution | |
44.5 to 47.9% (Si) | |
SiO2 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
silanedione | |
24261 | |
60.08 | |
Trace Metal Basis |
1610.0°C | |
Colorless to Yellow | |
5 g | |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
Solubility in water: insoluble | |
O=[Si]=O | |
60.08 | |
CHEBI:30563 | |
99.998% | |
Glass bottle |
Chemical Identifiers
14808-60-7 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
24261 | |
silanedione |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
CHEBI:30563 | |
O=[Si]=O |
Safety and Handling
GHS H Statement
May cause cancer by inhalation.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 238-878-4
RUO – Research Use Only