missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silicon(II) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
$1446.33 - $1446.33
Chemical Identifiers
CAS | 10097-28-6 |
---|---|
Molecular Formula | OSi |
Molecular Weight (g/mol) | 44.084 |
MDL Number | MFCD00151536 |
InChI Key | LIVNPJMFVYWSIS-UHFFFAOYSA-N |
Synonym | silicon monoxide, silylene, oxo, silicon ii oxide, unii-1oqn9cbg7l, 1oqn9cbg7l, silicon ii oxide, optical grade, sio, oxidosilicon, oxosilanylidene, oxosilylene |
PubChem CID | 66241 |
ChEBI | CHEBI:30588 |
IUPAC Name | oxoniumylidynesilanide |
SMILES | [O+]#[Si-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AA4171036
|
Thermo Scientific Chemicals
04171036 |
500 g |
Each for $1,446.33
|
|
||||||
Description
For vacuum deposition
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10097-28-6 | |
44.084 | |
LIVNPJMFVYWSIS-UHFFFAOYSA-N | |
66241 | |
oxoniumylidynesilanide |
OSi | |
MFCD00151536 | |
silicon monoxide, silylene, oxo, silicon ii oxide, unii-1oqn9cbg7l, 1oqn9cbg7l, silicon ii oxide, optical grade, sio, oxidosilicon, oxosilanylidene, oxosilylene | |
CHEBI:30588 | |
[O+]#[Si-] |
Specifications
1880°C | |
10097-28-6 | |
500 g | |
MFCD00151536 | |
LIVNPJMFVYWSIS-UHFFFAOYSA-N | |
oxoniumylidynesilanide | |
66241 | |
44.09 | |
Plastic bottle | |
2.13 |
>1702°C | |
Pieces | |
OSi | |
silicon monoxide, silylene, oxo, silicon ii oxide, unii-1oqn9cbg7l, 1oqn9cbg7l, silicon ii oxide, optical grade, sio, oxidosilicon, oxosilanylidene, oxosilylene | |
[O+]#[Si-] | |
44.084 | |
CHEBI:30588 | |
99.9% | |
2.13 | |
Silicon(II) oxide |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 233-232-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only