missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicon(IV) acetate, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01470318

 View more versions of this product

Catalog No. AA1470318


Explore available promotions

Description

Description

Silicon tetraacetate is used in the preparation of silicon dioxide thin films by a direct photochemical vapor deposition method. It serves as a precursor to prepare silicon complexes with monofunctional bidentate Schiff bases. It is also used as an alternative to silicon hydride and alkoxide for low-temperature silicon dioxide production. Further, it reacts with ethanol in the absence of water to get silica gel and ethyl acetate. In addition, it is employed as a sol-gel precursor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

160°C to 170°C (decomposition)
110°C
Crystalline
C8H12O8Si
UN3261
silicon tetraacetate, tetraacetoxysilane, tetraacetyl orthosilicate, unii-vz7lp47epp, vz7lp47epp, silicon iv acetate, anhydrous, acetic acid, 1,1',1,1'-tetraanhydride with silicic acid h4sio4, triacetoxysilyl acetate, silanetetrol, tetraacetate
YZVRVDPMGYFCGL-UHFFFAOYSA-N
triacetyloxysilyl acetate
68419
Moisture sensitive
Silicon(IV) acetate
562-90-3
50 g
MFCD00026184
14,8498
Soluble in acetone and benzene.
CC(=O)O[Si](OC(C)=O)(OC(C)=O)OC(C)=O
264.26
264.27

Chemical Identifiers

562-90-3
264.26
YZVRVDPMGYFCGL-UHFFFAOYSA-N
68419
CC(=O)O[Si](OC(C)=O)(OC(C)=O)OC(C)=O
C8H12O8Si
MFCD00026184
silicon tetraacetate, tetraacetoxysilane, tetraacetyl orthosilicate, unii-vz7lp47epp, vz7lp47epp, silicon iv acetate, anhydrous, acetic acid, 1,1',1,1'-tetraanhydride with silicic acid h4sio4, triacetoxysilyl acetate, silanetetrol, tetraacetate
triacetyloxysilyl acetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 209-239-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only