missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silicon(IV) chloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 428761000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silicon(IV) chloride | |
100 mL | |
SiCl4 | |
15, 8639 | |
Solubility in water: reacts violently | |
Cl[Si](Cl)(Cl)Cl | |
169.89 | |
169.9 | |
AcroSeal™ Glass bottle | |
1.34 |
10026-04-7,75-09-2 | |
Cl4Si | |
MFCD00011229 | |
silicon tetrachloride, silicon chloride, perchlorosilane, silane, tetrachloro, silicon iv chloride, tetrachlorosilicon, chlorid kremicity, silicio tetracloruro di, siliciumtetrachlorid, silicon chloride sicl4 | |
FDNAPBUWERUEDA-UHFFFAOYSA-N | |
tetrachlorosilane | |
24816 | |
1M, exact strength on the certificate of analysis | |
Clear colorless solution | |
1.3400g/mL |
RUO – Research Use Only