missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicon(IV) nitride, amorphous, nanopowder, 98.5+%, Thermo Scientific Chemicals

$88.51 - $884.33

Chemical Identifiers

CAS 12033-89-5
Molecular Formula N4Si3
Molecular Weight (g/mol) 140.28
MDL Number MFCD00011230
InChI Key HQVNEWCFYHHQES-UHFFFAOYSA-N
Synonym silicon nitride, silicon nitride si3n4, unii-qhb8t06idk, qhb8t06idk, silicon iv nitride, alpha phase, trisilicon tetranitride, nierite, nano silicon nitride, silicon iv nitride, silicon nitride, amorphous
PubChem CID 3084099
IUPAC Name Silicon nitride
SMILES N12[Si]34N5[Si]11N3[Si]25N41
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4561106
View Documents
Thermo Scientific Chemicals
04561106
5 g
Each for $88.51
 
AA4561114
View Documents
Thermo Scientific Chemicals
04561114
25 g
Each for $296.52
 
AA4561122
View Documents
Thermo Scientific Chemicals
04561122
100 g
Each for $884.33
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12033-89-5
140.28
HQVNEWCFYHHQES-UHFFFAOYSA-N
3084099
N12[Si]34N5[Si]11N3[Si]25N41
N4Si3
MFCD00011230
silicon nitride, silicon nitride si3n4, unii-qhb8t06idk, qhb8t06idk, silicon iv nitride, alpha phase, trisilicon tetranitride, nierite, nano silicon nitride, silicon iv nitride, silicon nitride, amorphous
Silicon nitride

Specifications

1900°C (sublimation)
12033-89-5
5 g
MFCD00011230
silicon nitride, silicon nitride si3n4, unii-qhb8t06idk, qhb8t06idk, silicon iv nitride, alpha phase, trisilicon tetranitride, nierite, nano silicon nitride, silicon iv nitride, silicon nitride, amorphous
N12[Si]34N5[Si]11N3[Si]25N41
140.28
140.28
Odorless
Silicon (IV) nitride, Amorphous
Gray
Powder
N4Si3
14,8497
HQVNEWCFYHHQES-UHFFFAOYSA-N
Silicon nitride
3084099
≥98.5%
3.2 g/mL
Safety and Handling

Safety and Handling

EINECSNumber : 234-796-8

RTECSNumber : VW0650000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only