Learn More
Silicon(IV) oxide, powder, 1.5 micron, 99.9%, Thermo Scientific Chemicals
$140.44 - $460.19
Chemical Identifiers
CAS | 7631-86-9 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.08 |
MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
Synonym | silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
IUPAC Name | dioxosilane |
SMILES | O=[Si]=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL1698714
|
Thermo Scientific Chemicals
L1698714 |
25 g |
N/A
|
|
||||||
AAL1698722
|
Thermo Scientific Chemicals
L1698722 |
100 g |
N/A
|
|
||||||
Description
Silicon(IV) oxide is used in a variety of applications such as manufacturing of glass, ceramics and abrasives, and water filtration. It is a common additive in dentifrice oral care applications and the production of foods, where it is used primarily as an anti-caking and free-flow agent in powdered foods, or to absorb water in hygroscopic applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSilicon(IV) oxide is used in a variety of applications such as manufacturing of glass, ceramics and abrasives, and water filtration. It is a common additive in dentifrice oral care applications and the production of foods, where it is used primarily as an anti-caking and free-flow agent in powdered foods, or to absorb water in hygroscopic applications.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with hydrogen fluoride.
Chemical Identifiers
7631-86-9 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
24261 | |
dioxosilane |
O2Si | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
CHEBI:30563 | |
O=[Si]=O |
Specifications
2230°C | |
7631-86-9 | |
25 g | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
60.09 | |
Odorless | |
Silicon(IV) oxide, 1.5 micron |
1710°C | |
Powder | |
O2Si | |
14,8493 | |
Insoluble in water. | |
O=[Si]=O | |
60.08 | |
CHEBI:30563 | |
99.9% | |
2 |
Safety and Handling
GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 231-545-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only