missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silicon Optical Window, 25.4mm (1.0 in.) dia. x 1mm (0.04 in.) thick, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 45380KS
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Silicon Optical Window | |
7440-21-3 | |
MFCD00085311 | |
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 | |
[Si] | |
28.09 | |
CHEBI:27573 |
25.4mm (1.0 in.) dia. x 1mm (0.04 in.) thick, Uncoated, Clear Aperture 85%, Surface Finsh 40/20 | |
Si | |
1 Ea. | |
XUIMIQQOPSSXEZ-UHFFFAOYSA-N | |
silicon | |
5461123 |
Chemical Identifiers
7440-21-3 | |
28.09 | |
XUIMIQQOPSSXEZ-UHFFFAOYSA-N | |
5461123 | |
silicon |
Si | |
MFCD00085311 | |
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 | |
CHEBI:27573 | |
[Si] |
Safety and Handling
EINECSNumber : 231-130-8
TSCA : Yes