missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicon plate, 12.5mm (0.5in) thick, 99.999% (metals basis), Thermo Scientific Chemicals

$586.21 - $2258.99

Chemical Identifiers

CAS 7440-21-3
Molecular Formula Si
Molecular Weight (g/mol) 28.09
MDL Number MFCD00085311
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
Synonym metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
PubChem CID 5461123
ChEBI CHEBI:27573
IUPAC Name silicon
SMILES [Si]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA39109FF
View Documents
Thermo Scientific Chemicals
039109FF
25 x 25 mm
Each for $586.21
 
AA39109FI
View Documents
Thermo Scientific Chemicals
039109FI
50 x 50 mm
Each for $723.16
 
AA39109GH
View Documents
Thermo Scientific Chemicals
039109GH
100 x 100 mm
Each for $2,258.99
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-21-3
28.09
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
5461123
silicon
Si
MFCD00085311
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
CHEBI:27573
[Si]

Specifications

7440-21-3
MFCD00085311
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
[Si]
28.09
CHEBI:27573
Odorless
Plate
(metals basis)
Si
25 x 25 mm
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
silicon
5461123
99.999%
≈19.09g/25x25mm
Silicon plate, 12.5mm (0.5 in.) thick
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-130-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only