missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silicon rod, 5.08cm (2.0 in.) dia., 99.999% (metals basis), Thermo Scientific™
Supplier: Thermo Scientific Chemicals 39103DH
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Silicon rod | |
7440-21-3 | |
Si | |
25 mm | |
XUIMIQQOPSSXEZ-UHFFFAOYSA-N | |
silicon | |
5461123 | |
99.999% | |
≈47.18g/cm |
5.08cm (2.0 in.) dia. | |
(metals basis) | |
MFCD00085311 | |
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 | |
[Si] | |
28.09 | |
CHEBI:27573 | |
Odorless | |
Rod |
Chemical Identifiers
7440-21-3 | |
28.09 | |
XUIMIQQOPSSXEZ-UHFFFAOYSA-N | |
5461123 | |
silicon |
Si | |
MFCD00085311 | |
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 | |
CHEBI:27573 | |
[Si] |
Safety and Handling
EINECSNumber : 231-130-8
TSCA : Yes
Recommended Storage : Ambient temperatures