missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicon slugs, 9.5mm (0.375 in.) dia. x 9.5mm (0.375 in.) length, 99.9999% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04392614

 View more versions of this product

Catalog No. AA4392614


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silicon slugs
7440-21-3
Si
25 g
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
silicon
5461123
99.9999%
≈1.57g/slug
9.5mm (0.375 in.) dia. x 9.5mm (0.375 in.) length
(metals basis)
MFCD00085311
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
[Si]
28.09
CHEBI:27573
Odorless
Slug

Chemical Identifiers

7440-21-3
28.09
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
5461123
silicon
Si
MFCD00085311
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
CHEBI:27573
[Si]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-130-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only