missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicone Oil, For oil baths up to 250°C, MilliporeSigma™

Supplier:  Technidata 1077421000

Catalog No. M1077421000


Explore available promotions

Description

Description

Specifications

Specifications

>250°C (1013hPa)
Colorless
Liquid
C16H22O2Si2
diphenyl-dimethylsiloxane copolymer, polydimethyl-diphenylsiloxane, viscosity 400cst.
CO[Si](C)(C)O[Si](C)(C1=CC=CC=C1)C2=CC=CC=C2
302.52
Plastic Bottle
1.04g/3 (20°C)
Sillicone
68083-14-7
1000 mL
315°C
ARWRSWALIGRKQA-UHFFFAOYSA-N
methoxy-dimethyl-[methyl(diphenyl)silyl]oxysilane
121233058
47 hPa (20°C)

Chemical Identifiers

68083-14-7
302.52
diphenyl-dimethylsiloxane copolymer, polydimethyl-diphenylsiloxane, viscosity 400cst.
methoxy-dimethyl-[methyl(diphenyl)silyl]oxysilane
C16H22O2Si2
ARWRSWALIGRKQA-UHFFFAOYSA-N
121233058
CO[Si](C)(C)O[Si](C)(C1=CC=CC=C1)C2=CC=CC=C2
Safety and Handling

Safety and Handling

RTECSNumber : VW3146875

SDS
Documents

Documents

Promotions

Promotions