missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silicone oil, for melting point and boiling point apparatuses, Thermo Scientific Chemicals
$150.00 - $468.55
Chemical Identifiers
CAS | 63148-62-9 |
---|---|
Molecular Formula | (C2H6OSi)n |
Molecular Weight (g/mol) | NaN |
MDL Number | MFCD00132673 |
Synonym | Poly(dimethylsiloxane) |
IUPAC Name | Polydimethylsiloxane |
SMILES | C[Si](C)(-*)O-* |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC163855000
|
Thermo Scientific Chemicals
163855000 |
500 g | Glass bottle |
Each for $150.00
|
|
||||
AC163850010
|
Thermo Scientific Chemicals
163850010 |
1 kg | Glass bottle |
Each for $195.63
|
|
||||
AC163850025
|
Thermo Scientific Chemicals
163850025 |
2.5 kg | Glass bottle |
Each for $468.55
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
63148-62-9 | |
NaN | |
Poly(dimethylsiloxane) | |
C[Si](C)(-*)O-* |
(C2H6OSi)n | |
MFCD00132673 | |
Polydimethylsiloxane |
Specifications
-50.0°C | |
63148-62-9 | |
500 g | |
MFCD00132673 | |
Poly(dimethylsiloxane) | |
C[Si](C)(-*)O-* | |
NaN | |
Glass bottle | |
0.96 | |
Silicone oil, For melting point and boiling point apparatuses |
Colorless | |
Oily Liquid | |
(C2H6OSi)n | |
300°C | |
Solubility in water: practically insoluble | |
Polydimethylsiloxane | |
Authentic | |
1.4030 to 1.4060 | |
0.9600g/mL |
RUO – Research Use Only