Learn More
Silver bromide, 99.5%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01142522
Description
Silver bromide is used as the light-sensitive component on ordinary photographic film and plates and also used in I.R. applications and ionic semiconductors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver bromide | |
7785-23-1 | |
Powder | |
99.9% | |
MFCD00003398 | |
Insoluble in water,alcohol and acids. Partially soluble in ammonia. | |
Br[Ag] | |
187.772 | |
187.78 | |
Light sensitive | |
6.47 |
432°C | |
Yellow | |
100 g | |
AgBr | |
silver bromide, bromargyrite, bromyrite, nanoparticles, silver i bromide, silver bromide, premion | |
ADZWSOLPGZMUMY-UHFFFAOYSA-M | |
bromosilver | |
66199 | |
Odorless | |
Pale yellow powder/flake/crystalline/beads |
Chemical Identifiers
7785-23-1 | |
187.772 | |
ADZWSOLPGZMUMY-UHFFFAOYSA-M | |
66199 | |
Br[Ag] |
AgBr | |
MFCD00003398 | |
silver bromide, bromargyrite, bromyrite, nanoparticles, silver i bromide, silver bromide, premion | |
bromosilver |
Safety and Handling
Use appropriate containment to avoid environmental contamination. This material and/or its container must be disposed of as hazardous waste.
H400-H410: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
DOTInformation : DOT Class: 9, Packaging Group: III
EINECSNumber : 232-076-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only