Learn More
Silver chloride, 99+%, extra pure, Thermo Scientific Chemicals
$148.76 - $1,491.00
Chemical Identifiers
CAS | 7783-90-6 |
---|---|
Molecular Formula | AgCl |
Molecular Weight (g/mol) | 143.32 |
MDL Number | MFCD00003399 |
InChI Key | HKZLPVFGJNLROG-UHFFFAOYSA-M |
IUPAC Name | silver(1+) chloride |
SMILES | [Cl-].[Ag+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC211270100
|
Thermo Scientific Chemicals
211270100 |
10 g | Glass bottle |
N/A
|
|
||||
AC211270500
|
Thermo Scientific Chemicals
211270500 |
50 g | Glass bottle |
N/A
|
|
||||
AC211272500
|
Thermo Scientific Chemicals
211272500 |
250 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7783-90-6 | |
143.32 | |
HKZLPVFGJNLROG-UHFFFAOYSA-M | |
[Cl-].[Ag+] |
AgCl | |
MFCD00003399 | |
silver(1+) chloride |
Specifications
1550.0°C | |
7783-90-6 | |
10 g | |
AgCl | |
MFCD00003399 | |
15,8648 | |
HKZLPVFGJNLROG-UHFFFAOYSA-M | |
silver(1+) chloride | |
143.32 | |
Extra Pure | |
Silver chloride |
455.0°C | |
Powder | |
99+% | |
AgCl | |
05,580; 10,347 | |
Solubility in water: 1.93 mg/l (25°C). Other solubilities: insoluble in alcohol | |
[Cl-].[Ag+] | |
143.32 | |
≥99% | |
Glass bottle |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Absorb spillage to prevent material damage.
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.
GHS Signal Word: Warning
EINECSNumber : 232-033-3
RUO – Research Use Only