Learn More
Silver cyanate, 98%, Thermo Scientific Chemicals
$130.97 - $646.00
Chemical Identifiers
CAS | 3315-16-0 |
---|---|
Molecular Formula | CHAgNO |
Molecular Weight (g/mol) | 150.893 |
MDL Number | MFCD00011512 |
InChI Key | VAQVDZZAAMAZAO-UHFFFAOYSA-N |
Synonym | silver,cyanate, cyanic acid; silver |
PubChem CID | 45051745 |
IUPAC Name | cyanic acid;silver |
SMILES | C(#N)O.[Ag] |
Description
Silver cyanate is a chemosterilant in adult house flies. It is also used as a reagent to prepare 1-(3-thianyl)uracil and 9-(3-thianyl)adenine derivatives, which has potential anti-human immunodeficiency virus (HIV) and antitumour activity.It is used as a silver plating solution in the electroplating process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3315-16-0 | |
150.893 | |
VAQVDZZAAMAZAO-UHFFFAOYSA-N | |
45051745 | |
C(#N)O.[Ag] |
CHAgNO | |
MFCD00011512 | |
silver,cyanate, cyanic acid; silver | |
cyanic acid;silver |
Specifications
3315-16-0 | |
Powder | |
98% | |
MFCD00011512 | |
silver,cyanate, cyanic acid; silver | |
VAQVDZZAAMAZAO-UHFFFAOYSA-N | |
cyanic acid;silver | |
45051745 | |
Odorless | |
4 |
White | |
5 g | |
CHAgNO | |
3655040 | |
Soluble in ammonia,nitric acid,potassium cyanide,ammonium hydroxide. Insoluble in alcohol and dilute acids. | |
C(#N)O.[Ag] | |
150.893 | |
149.89 | |
Light sensitive | |
Silver cyanate |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : DOT Class: 9, Packaging Group: III
EINECSNumber : 222-006-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only