Learn More
Silver cyanide, 99%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 180230500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver cyanide | |
99% | |
White | |
50 g | |
CAgN | |
MFCD00003409 | |
15, 8651 | |
Solubility in water: insoluble. Other solubilities: sol. in nh4oh,alkali cyanides,boiling conc. hno3,insoluble in alcohol and diluted acids | |
[C-]#N.[Ag+] | |
133.89 | |
133.89 | |
Analytical |
>300°C | |
506-64-9 | |
Powder | |
98.5% min | |
AgCN | |
01,1006; 05,581; 07,320 | |
silver cyanide, silver cyanide ag cn, silver 1+ cyanide, kyanid stribrny czech, cyanure d'argent french, rcra waste number p104, silver i cyanide, rcra waste no. p104, kyanid stribrny, cyanure d'argent | |
LFAGQMCIGQNPJG-UHFFFAOYSA-N | |
silver;cyanide | |
10475 | |
99% | |
Glass bottle |
Chemical Identifiers
506-64-9 | |
133.89 | |
LFAGQMCIGQNPJG-UHFFFAOYSA-N | |
10475 | |
[C-]#N.[Ag+] |
CAgN | |
MFCD00003409 | |
silver cyanide, silver cyanide ag cn, silver 1+ cyanide, kyanid stribrny czech, cyanure d'argent french, rcra waste number p104, silver i cyanide, rcra waste no. p104, kyanid stribrny, cyanure d'argent | |
silver;cyanide |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Fatal if swallowed.
Fatal in contact with skin.
Contact with acids liberates very toxic gas.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently
GHS Signal Word: Danger
EINECSNumber : 208-048-6
RTECSNumber : VW3850000
TSCA : TSCA
RUO – Research Use Only