missing translation for 'onlineSavingsMsg'
Learn More

Silver(I) oxide, 99+%

CAS: 20667-12-3 | Ag2O | 231.74 g/mol

Supplier:  Thermo Scientific Chemicals 205800100

 View more versions of this product

Catalog No. AC205800100


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Specifications

Silver(I) oxide
20667-12-3
10 g
Ag2O
MFCD00003404
15, 8660
KHJDQHIZCZTCAE-UHFFFAOYSA-N
(argentiooxy)silver
231.74
Glass bottle
Black
Powder
99+%
Ag2O
01,1011; 02,368; 03,252; 04,430; 05,583; 06,515; 07,321; 08,442; 10,350; 11,468
Solubility in water: slightly soluble. Other solubilities: freely soluble in dil.hno3 and ammonia,practically insoluble in alcohol
[Ag]O[Ag]
231.74
≥99%

Chemical Identifiers

20667-12-3
231.74
KHJDQHIZCZTCAE-UHFFFAOYSA-N
[Ag]O[Ag]
Ag2O
MFCD00003404
(argentiooxy)silver
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
May cause fire or explosion; strong oxidizer.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 243-957-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only