missing translation for 'onlineSavingsMsg'
Learn More

Silver iodide, 99%, pure

CAS: 7783-96-2 | AgI | 234.77 g/mol

$97.67 - $516.94

Chemical Identifiers

CAS 7783-96-2
Molecular Formula AgI
Molecular Weight (g/mol) 234.77
MDL Number MFCD00003412
InChI Key MSFPLIAKTHOCQP-UHFFFAOYSA-M
Synonym silver iodide, neosiluol, neosilvol, silver monoiodide, silver 1+ iodide, colloidal silver iodide, iodanylsilver, silver iodide metals basis, ccris 8490
PubChem CID 24563
IUPAC Name iodosilver
SMILES [Ag]I
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC210420050
SDS View Documents
Thermo Scientific Chemicals
210420050
5 g Glass bottle
Each for $97.67
Only null left
 
AC210420250
SDS View Documents
Thermo Scientific Chemicals
210420250
25 g Glass bottle
Each for $293.71
Only null left
 
AC210421000
SDS View Documents
Thermo Scientific Chemicals
210421000
100 g Glass bottle
Each for $516.94
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7783-96-2
234.77
MSFPLIAKTHOCQP-UHFFFAOYSA-M
24563
[Ag]I
AgI
MFCD00003412
silver iodide, neosiluol, neosilvol, silver monoiodide, silver 1+ iodide, colloidal silver iodide, iodanylsilver, silver iodide metals basis, ccris 8490
iodosilver

Specifications

1506°C
7783-96-2
Granular Powder
98.5% min. (ex Ag)
AgI
15, 8655
Solubility in water: 0.03mg/l
[Ag]I
234.77
234.77
Pure
5.68
Silver iodide, Pure
557°C
Yellow
5 g
AgI
MFCD00003412
silver iodide, neosiluol, neosilvol, silver monoiodide, silver 1+ iodide, colloidal silver iodide, iodanylsilver, silver iodide metals basis, ccris 8490
MSFPLIAKTHOCQP-UHFFFAOYSA-M
iodosilver
24563
99%
Glass bottle
5.6800g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.

GHS Signal Word: Warning

EINECSNumber : 232-038-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only