Learn More
Silver nitrate, ACS, 99.9+% (metals basis), Thermo Scientific Chemicals
Silver nitrate, AgNO3, CAS Number-7761-88-8, silvernitrate, silver i nitrate, argenti nitras, nitric acid silver i salt, nitrate d'argent, lunar caustic, argerol, silver mononitrate, silver nitrate, silbernitrat, 100g, >99.9% (metals basis), CHEBI:32130, 4.352 g/mL
$69.62 - $2,292.27
Chemical Identifiers
CAS | 7761-88-8 |
---|---|
Molecular Formula | AgNO3 |
Molecular Weight (g/mol) | 169.87 |
MDL Number | MFCD00003414 |
InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
Synonym | silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol |
PubChem CID | 24470 |
ChEBI | CHEBI:32130 |
IUPAC Name | silver(1+) nitrate |
SMILES | [Ag+].[O-][N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1141406
|
Thermo Scientific Chemicals
01141406 |
5 g |
N/A
|
|
|||||
AA1141414
|
Thermo Scientific Chemicals
01141414 |
25 g |
N/A
|
|
|||||
AA1141422
|
Thermo Scientific Chemicals
01141422 |
100 g |
N/A
|
|
|||||
AA1141436
|
Thermo Scientific Chemicals
01141436 |
500 g |
N/A
|
|
|||||
Description
Silver nitrate is employed in Silver plating, photography, manufacturing of other silver compounds, mirrors, coloring porcelain, and staining in histology. It finds use as a stain in scanning electron microscopy, and a key ingredient in several silver-based antiseptic and antibacterial pharmaceutical compositions. Silver(I) nitrate is used to separate mixtures of alkenes by selective absorption, and for deprotection and oxidation reactions in organic synthesis. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7761-88-8 | |
169.87 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
24470 | |
silver(1+) nitrate |
AgNO3 | |
MFCD00003414 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
CHEBI:32130 | |
[Ag+].[O-][N+]([O-])=O |
Specifications
212°C | |
Crystalline | |
>99.9% (metals basis) | |
MFCD00003414 | |
Freely soluble in water,alcohol and aqueous ammonia. Slightly soluble in ether. | |
[Ag+].[O-][N+]([O-])=O | |
169.87 | |
CHEBI:32130 | |
ACS Reagent | |
4.352 g/mL |
7761-88-8 | |
5 g | |
AgNO3 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
silver(1+) nitrate | |
24470 | |
169.87 | |
Light sensitive | |
Silver nitrate, For AAS analysis |
Safety and Handling
P220-P221-P234-P260-P262-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H272-H290-H314-H335-H373
DOTInformation : Hazard Class: 5.1; Packaging Group: II
EINECSNumber : 231-853-9
RTECSNumber : VW4725000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only