missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver Nitrate, Certified, 0.1000N ±0.0005N (0.1M), LabChem™

Supplier:  LabChem LC227001

 View more versions of this product

Catalog No. LC227001


Explore available promotions

Description

Description

Specifications

Specifications

Silver Nitrate
Colorless
Liquid
AgNO3
MFCD00003414
Soluble in water
[Ag+].[O-][N+]([O-])=O
169.87
CHEBI:32130
Certified
Passes Test
1g/mL
1g/mL
7761-88-8,7732-18-5
98.3,1.7
500 mL
AgNO3
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
24470
169.87
0.1000N ±0.0005N (0.1M)
Amber Glass
Nitrogen oxides; Fume; Carbon monoxide; Carbon dioxide

Chemical Identifiers

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver(1+) nitrate
AgNO3
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130
[Ag+].[O-][N+]([O-])=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin and serious eye irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.

Warning

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions