Learn More
Silver Nitrate, Certified, 0.1000N ±0.0005N (0.1M), LabChem™
Supplier: LabChem LC227006
Specifications
Silver Nitrate | |
Colorless | |
Liquid | |
AgNO3 | |
MFCD00003414 | |
Soluble in water | |
[Ag+].[O-][N+]([O-])=O | |
169.87 | |
CHEBI:32130 | |
Certified | |
Passes Test | |
1g/mL | |
1g/mL |
7761-88-8,7732-18-5 | |
98.3,1.7 | |
10 L | |
AgNO3 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
silver(1+) nitrate | |
24470 | |
169.87 | |
0.1000N ±0.0005N (0.1M) | |
Cubitainer™ | |
Nitrogen oxides; Fume; Carbon monoxide; Carbon dioxide |
Chemical Identifiers
7761-88-8 | |
169.87 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
24470 | |
silver(1+) nitrate |
AgNO3 | |
MFCD00003414 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
CHEBI:32130 | |
[Ag+].[O-][N+]([O-])=O |
Safety and Handling
GHS H Statement
Causes skin and serious eye irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
Warning
Recommended Storage : Room Temperature