missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silver Nitrate (Crystalline/Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical FLS181100
Specifications
444°C | |
212°C | |
White | |
Solid | |
Assay: ≥99.7% | |
MFCD00003414 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
silver(1+) nitrate | |
24470 | |
169.87 | |
Certified ACS | |
Glass Bottle |
Silver Nitrate | |
7761-88-8 | |
5.4 to 6.4 | |
100 g | |
AgNO3 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
[Ag+].[O-][N+]([O-])=O | |
169.87 | |
CHEBI:32130 | |
≥99.7% | |
Pass Test |
Chemical Identifiers
7761-88-8 | |
169.87 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
24470 | |
silver(1+) nitrate |
AgNO3 | |
MFCD00003414 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
CHEBI:32130 | |
[Ag+].[O-][N+]([O-])=O |
Safety and Handling
DOTInformation : DOT Class 5.1: Oxidizer