missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver Nitrate (Crystalline/Certified ACS), Fisher Chemical™

Supplier:  Fisher Chemical FLS18125

 View more versions of this product

Catalog No. S181-25



Explore available promotions

Description

Description

Specifications

Specifications

444°C
212°C
White
Solid
Assay: ≥99.7%
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
24470
169.87
Certified ACS
Glass Bottle
Silver Nitrate
7761-88-8
5.4 to 6.4
25 g
AgNO3
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
[Ag+].[O-][N+]([O-])=O
169.87
CHEBI:32130
≥99.7%
Pass Test

Chemical Identifiers

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver(1+) nitrate
AgNO3
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130
[Ag+].[O-][N+]([O-])=O
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 5.1: Oxidizer

SDS Documents

Documents

Promotions

Promotions