missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver nitrite, 99% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01141809

 View more versions of this product

Catalog No. AA1141809


Explore available promotions

Description

Description

Silver nitrite is used in the silver plating process, hair dye, inks, permanent marker pens and determination of chloride ions in water and drilling fluids. It is used as Tollen’s reagent and used in the formation of aniline and nitro compounds in victor mayer reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silver nitrite
7783-99-5
Crystalline
99% (metals basis)
MFCD00003413
Soluble in hot water.Insoluble in ethanol.
N(=O)[O-].[Ag+]
153.873
153.88
Light sensitive
4.45 g/cm3
140°C (decomposition)
White
10 g
AgNO2
silver nitrite, unii-t3mz57ogif, t3mz57ogif, silbernitrit, nitrous acid, silver 1+ salt, silver i nitrite, silvernitrite, acmc-20ajq7, ag.no2, silver nitrite 10g
KKKDGYXNGYJJRX-UHFFFAOYSA-M
silver;nitrite
160904
Odorless
Light yellow crystals

Chemical Identifiers

7783-99-5
153.873
KKKDGYXNGYJJRX-UHFFFAOYSA-M
160904
N(=O)[O-].[Ag+]
AgNO2
MFCD00003413
silver nitrite, unii-t3mz57ogif, t3mz57ogif, silbernitrit, nitrous acid, silver 1+ salt, silver i nitrite, silvernitrite, acmc-20ajq7, ag.no2, silver nitrite 10g
silver;nitrite
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P370+P378q-P501c

H272-H302-H315-H319

DOTInformation : DOT Class: 5.1, Packaging Group: II

EINECSNumber : 232-041-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only