Learn More
Silver p-toluenesulfonate, 98+%, Thermo Scientific Chemicals
$111.03 - $328.71
Chemical Identifiers
CAS | 16836-95-6 |
---|---|
Molecular Formula | C7H8AgO3S |
Molecular Weight (g/mol) | 280.07 |
MDL Number | MFCD00007511 |
InChI Key | BRYKBDMLJJLFAB-UHFFFAOYSA-N |
Synonym | silver p-toluenesulfonate, silver tosylate, silverp-toluenesulfonate, silver p-toluenesulphonate, silver 1+ tosylate, silver p-tosylate, benzenesulfonic acid, 4-methyl-, silver 1+ salt, p-toluenesulfonic acid silver salt, silver i 4-methylbenzenesulfonate, silver p-methylbenzenesulfonate |
PubChem CID | 85606 |
IUPAC Name | silver;4-methylbenzenesulfonate |
SMILES | [Ag].CC1=CC=C(C=C1)S(O)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0030706
|
Thermo Scientific Chemicals
L0030706 |
5 g |
Each for $111.03
|
|
||||||
AAL0030714
|
Thermo Scientific Chemicals
L0030714 |
25 g |
Each for $328.71
|
|
||||||
Description
Silver p-toluenesulfonate is used to converts alkyl halides to tosylates, and benzyl selenyl chlorides into their corresponding selenyl tosylates. It used in promotion of the leaving ability of halogens. For use in the conversion of alkyl halides to alkyl tosylates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSilver p-toluenesulfonate is used to converts alkyl halides to tosylates, and benzyl selenyl chlorides into their corresponding selenyl tosylates. It used in promotion of the leaving ability of halogens. For use in the conversion of alkyl halides to alkyl tosylates.
Solubility
Soluble in water.
Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
16836-95-6 | |
280.07 | |
BRYKBDMLJJLFAB-UHFFFAOYSA-N | |
85606 | |
[Ag].CC1=CC=C(C=C1)S(O)(=O)=O |
C7H8AgO3S | |
MFCD00007511 | |
silver p-toluenesulfonate, silver tosylate, silverp-toluenesulfonate, silver p-toluenesulphonate, silver 1+ tosylate, silver p-tosylate, benzenesulfonic acid, 4-methyl-, silver 1+ salt, p-toluenesulfonic acid silver salt, silver i 4-methylbenzenesulfonate, silver p-methylbenzenesulfonate | |
silver;4-methylbenzenesulfonate |
Specifications
16836-95-6 | |
C7H8AgO3S | |
UN3077 | |
Soluble in water. | |
[Ag].CC1=CC=C(C=C1)S(O)(=O)=O | |
280.07 | |
279.07 | |
Odorless | |
Silver p-toluenesulfonate |
5 g | |
MFCD00007511 | |
silver p-toluenesulfonate, silver tosylate, silverp-toluenesulfonate, silver p-toluenesulphonate, silver 1+ tosylate, silver p-tosylate, benzenesulfonic acid, 4-methyl-, silver 1+ salt, p-toluenesulfonic acid silver salt, silver i 4-methylbenzenesulfonate, silver p-methylbenzenesulfonate | |
BRYKBDMLJJLFAB-UHFFFAOYSA-N | |
silver;4-methylbenzenesulfonate | |
85606 | |
≥98% | |
Light sensitive |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 240-859-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only