Learn More
Silver phosphate, 99% (metals basis), Thermo Scientific Chemicals
$50.96 - $166.73
Chemical Identifiers
CAS | 7784-09-0 |
---|---|
Molecular Formula | Ag3O4P |
Molecular Weight (g/mol) | 418.57 |
MDL Number | MFCD00014148 |
InChI Key | FJOLTQXXWSRAIX-UHFFFAOYSA-K |
Synonym | trisilver phosphate, trisilver 1+ phosphate, unii-zl6t4y1xp8, zl6t4y1xp8, silver orthophosphate, trisilver orthophosphate, silver phosphate ag3po4, silberphosphat, trisilver 1+ orthophosphate, silver orthophosphate ag3po4 |
PubChem CID | 159913 |
IUPAC Name | trisilver;phosphate |
SMILES | [Ag+].[Ag+].[Ag+].[O-]P([O-])([O-])=O |
Description
Silver phosphate finds applications in photography, and in silver staining of biological materials. It is used as a photocatalyst for the visible light photochemical splitting of water, and for production of activated oxygen by the same method. It is employed in Crystal growth for scintillation, optics and dosimetry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7784-09-0 | |
418.57 | |
FJOLTQXXWSRAIX-UHFFFAOYSA-K | |
159913 | |
[Ag+].[Ag+].[Ag+].[O-]P([O-])([O-])=O |
Ag3O4P | |
MFCD00014148 | |
trisilver phosphate, trisilver 1+ phosphate, unii-zl6t4y1xp8, zl6t4y1xp8, silver orthophosphate, trisilver orthophosphate, silver phosphate ag3po4, silberphosphat, trisilver 1+ orthophosphate, silver orthophosphate ag3po4 | |
trisilver;phosphate |
Specifications
849°C | |
Gray to Black | |
5 g | |
Ag3O4P | |
trisilver phosphate, trisilver 1+ phosphate, unii-zl6t4y1xp8, zl6t4y1xp8, silver orthophosphate, trisilver orthophosphate, silver phosphate ag3po4, silberphosphat, trisilver 1+ orthophosphate, silver orthophosphate ag3po4 | |
FJOLTQXXWSRAIX-UHFFFAOYSA-K | |
trisilver;phosphate | |
159913 | |
Odorless | |
Yellow powder | |
Silver phosphate |
7784-09-0 | |
Powder | |
99% | |
MFCD00014148 | |
Practically insoluble in water. Slightly soluble in dilute acetic acid. Freely soluble in dilute HNO3,ammonia,ammonium carbonate,alkali cyanides and thiosulfates. | |
[Ag+].[Ag+].[Ag+].[O-]P([O-])([O-])=O | |
418.57 | |
418.58 | |
Light sensitive | |
6.37 |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : DOT Class: 9, Packaging Group: III
EINECSNumber : 232-049-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only