missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver rod, 6.35mm (0.25 in.) dia., 99.9% (metals basis), Thermo Scientific Chemicals

$87.66 - $1508.42

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA12188BP
View Documents
Thermo Scientific Chemicals
12188BP
2 cm
Each for $87.66
 
AA12188BS
View Documents
Thermo Scientific Chemicals
012188BS
10 cm
Each for $352.45
 
AA12188BW
View Documents
Thermo Scientific Chemicals
012188BW
50 cm
Each for $1,508.42
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-22-4
107.87
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]

Specifications

7440-22-4
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
[Ag]
107.87
CHEBI:9141
Characteristic
Rod
(metals basis)
Ag
2 cm
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
23954
99.9%
≈3.32g/cm
Silver rod, 6.35mm (0.25 in.) dia.
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-131-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only