missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Silver shot, 1-7mm (0.08-0.12in), Premion™, 99.999% (metals basis)

$209.70 - $3146.14

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
IUPAC Name silver
SMILES [Ag]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4739809
View Documents
Thermo Scientific™
4739809
10 g
Each for $209.70
 
AA4739818
View Documents
Thermo Scientific™
4739818
50 g
Each for $789.30
 
AA4739830
View Documents
Thermo Scientific™
4739830
250 g
Each for $3,146.14
 
Description

Description

Specifications

Chemical Identifiers

7440-22-4
107.87
BQCADISMDOOEFD-UHFFFAOYSA-N
[Ag]
Ag
MFCD00003397
silver

Specifications

7440-22-4
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
99.999
Ag
10 g
[Ag]
107.87
Solid
Videos
Safety and Handling

Safety and Handling

ShelfLife : 30 Years

EINECSNumber : 231-131-3

SDS
missing translation for 'documents'

missing translation for 'documents'