missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver sulfate, pure, 10 g/l solution in sulfuric acid
CAS: 10294-26-5 | Ag2H2O4S | 313.81 g/mol
$264.70 - $659.01
Specifications
| Chemical Name or Material | Silver sulfate |
|---|---|
| Color | Colorless |
| Name Note | 10g/L Solution in Sulfuric Acid |
| CAS | 10294-26-5,7664-93-9 |
| Molecular Formula | Ag2H2O4S |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC279480010
|
Thermo Scientific Chemicals
279480010 |
1 L |
Each for $264.70
|
|
|||||
|
AC279480025
|
Thermo Scientific Chemicals
279480025 |
2.5 L |
Each for $659.01
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Silver sulfate | |
| 10g/L Solution in Sulfuric Acid | |
| Ag2H2O4S | |
| MFCD00003407 | |
| 1.8400g/mL | |
| silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt | |
| [Ag+].[Ag+].OS(O)(=O)=O | |
| 313.81 | |
| 311.79 | |
| Glass bottle | |
| Liquid |
| Colorless | |
| 10294-26-5,7664-93-9 | |
| Ag2SO4 | |
| 01,1015; 03,254; 04,435 | |
| 15, 8668 | |
| YPNVIBVEFVRZPJ-UHFFFAOYSA-N | |
| disilver(1+) sulfuric acid | |
| 159865 | |
| Pure | |
| 1.84 |
RUO – Research Use Only