Learn More
Silver tetrafluoroborate, 99%, Thermo Scientific Chemicals
Light Sensitive and Hygroscopic
Supplier: Thermo Scientific Chemicals 01153918
Description
Silver tetrafluoroborate finds applications as a useful reagent in inorganic, organic and organomettalic chemistry. It acts as a moderately strong oxidant in dichloromethane, replaces halide anions and ligands with weakly coordinating tetrafluoroborate anions, and behaves as a catalyst for cyclization reactions. It is a potent promoter for chemical glycosylation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver tetrafluoroborate | |
14104-20-2 | |
Crystalline | |
99% | |
MFCD00003408 | |
Soluble in water and polar organic solvents. | |
[Ag+].F[B-](F)(F)F | |
194.67 | |
194.67 | |
Hygroscopic; Light sensitive |
200°C (decomposition) | |
Brown to White | |
50 g | |
AgBF4 | |
silver tetrafluoroborate, silver borofluoride, silver fluoborate, silver fluoroborate, silver i tetrafluoroborate, borate 1-, tetrafluoro-, silver 1+, silver tetrafluroborate, acmc-209clm | |
CCAVYRRHZLAMDJ-UHFFFAOYSA-N | |
silver;tetrafluoroborate | |
159722 | |
Odorless | |
White to pale brown crystals or powder |
Chemical Identifiers
14104-20-2 | |
194.67 | |
CCAVYRRHZLAMDJ-UHFFFAOYSA-N | |
159722 | |
[Ag+].F[B-](F)(F)F |
AgBF4 | |
MFCD00003408 | |
silver tetrafluoroborate, silver borofluoride, silver fluoborate, silver fluoroborate, silver i tetrafluoroborate, borate 1-, tetrafluoro-, silver 1+, silver tetrafluroborate, acmc-209clm | |
silver;tetrafluoroborate |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : DOT Class: 8, Packaging Group: II
EINECSNumber : 237-956-5
RTECSNumber : ED2875000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only