Learn More
Silver trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals
$47.66 - $499.71
Chemical Identifiers
CAS | 2923-28-6 |
---|---|
Molecular Formula | CAgF3O3S |
Molecular Weight (g/mol) | 256.93 |
MDL Number | MFCD00013226 |
InChI Key | QRUBYZBWAOOHSV-UHFFFAOYSA-M |
Synonym | silver trifluoromethanesulfonate, silver triflate, agotf, silver trifluoromethanesulphonate, trifluoromethanesulfonic acid silver salt, silver i trifluoromethanesulfonate, silver trifluoromethylsulfonate, methanesulfonic acid, trifluoro-, silver 1+ salt, silver i trifluoromethanesulphonate, silver 1+ ion triflate |
PubChem CID | 76223 |
IUPAC Name | silver;trifluoromethanesulfonate |
SMILES | [Ag+].[O-]S(=O)(=O)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA8872204
|
Thermo Scientific Chemicals
08872204 |
2 g |
Each for $47.66
|
|
|||||
AA8872209
|
Thermo Scientific Chemicals
08872209 |
10 g |
Each for $181.52
|
|
|||||
AA8872218
|
Thermo Scientific Chemicals
08872218 |
50 g |
Each for $499.71
|
|
|||||
Description
Silver trifluoromethanesulfonate is useful in halide abstraction. It helps in activating sulphur protecting groups at the anomeric centres of carbohydrates in the presence of iodine. It is employed as a triflating agent in combination with Palladium compounds, as a catalyst for cross coupling reactions, and in several organic transformation reactions. Polyurethanes prepared from silver triflate exhibit high permeability and selectivity towards olefins in their separation from paraffin mixtures. Polyethylene oxide - silver triflate composites find application in humidity sensors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2923-28-6 | |
256.93 | |
QRUBYZBWAOOHSV-UHFFFAOYSA-M | |
76223 | |
[Ag+].[O-]S(=O)(=O)C(F)(F)F |
CAgF3O3S | |
MFCD00013226 | |
silver trifluoromethanesulfonate, silver triflate, agotf, silver trifluoromethanesulphonate, trifluoromethanesulfonic acid silver salt, silver i trifluoromethanesulfonate, silver trifluoromethylsulfonate, methanesulfonic acid, trifluoro-, silver 1+ salt, silver i trifluoromethanesulphonate, silver 1+ ion triflate | |
silver;trifluoromethanesulfonate |
Specifications
356°C | |
White | |
2 g | |
CAgF3O3S | |
EM>10, | |
Soluble in water and also in most organic solvents. | |
[Ag+].[O-]S(=O)(=O)C(F)(F)F | |
256.93 | |
256.93 | |
Light sensitive |
2923-28-6 | |
Crystalline | |
98% | |
MFCD00013226 | |
silver trifluoromethanesulfonate, silver triflate, agotf, silver trifluoromethanesulphonate, trifluoromethanesulfonic acid silver salt, silver i trifluoromethanesulfonate, silver trifluoromethylsulfonate, methanesulfonic acid, trifluoro-, silver 1+ salt, silver i trifluoromethanesulphonate, silver 1+ ion triflate | |
QRUBYZBWAOOHSV-UHFFFAOYSA-M | |
silver;trifluoromethanesulfonate | |
76223 | |
Odorless | |
Silver trifluoromethanesulfonate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : DOT Class: 8, Packaging Group: III
EINECSNumber : 220-882-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only