missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver wire, 0.5mm (0.02in) dia, 1/2 hard, 99.9% (metals basis), Thermo Scientific Chemicals

Form: Approx. 2.06g/m, Temper: 1/2 hard

Supplier:  Thermo Scientific Chemicals 011433H2

 View more versions of this product

Catalog No. AA11433H2


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silver
1/2 Hard
Silver-colored wire
Ag
50 m
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
23954
Characteristic
Silver
7440-22-4
99.9% (Metals basis)
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
[Ag]
107.87
CHEBI:9141
Wire

Chemical Identifiers

7440-22-4
107.87
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-131-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only