Honeywell™ SilverShield™ SSG Gloves
Unique five-layer fabric is resistant to over 280 different chemicals including alcohols, aliphatics, aromatice, chlorines, ketones and esters.
Supplier:  Honeywell™ SSG2910
 View more versions of this product
Catalog No. 19-150-0892
Explore available promotions
Description
Description
  • Unique five-layer construction provides chemical protection
  • Ideal for hospitals, agriculture, military and hazardous material handling
  • Easily worn under heavy-duty outer gloves
  • Use as a primary glove where there is minimal risk of tearing or puncturing
  • Resistant to over 280 different chemicals, including alcohols, aliphatics, aromatics, chlorines, ketones and esters
  • Does not contain latex or chemical accelerators
Specifications
Specifications
OSHA 29 CFR 1915.157
Silver
Yes
10
0.06 mm
Bacteria, Fungi only
Smooth
SSG29/10
29 in.
EN 420, EN 374-3
Ambidextrous
Polyethylene, EVA
2.7 mil
EN 388
Straight
Unlined
SilverShield™ SSG
74 cm