missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific Capinsert™ For Cryovial Tubes

The Capinsert™ For Cryovial Tubes inserts fit precisely into the cap of the Cryovial™ for color identification.

$32.68 - $36.69

Specifications

Material Polypropylene
For Use With (Equipment) Cryovial™ Tubes
View More Specs

Products 8
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
22-038-503
View Documents
Simport™ Scientific
T31211
Burnt Orange
Case of 500 Each for $34.66
Only null left
 
22-038-501
View Documents
Simport™ Scientific
T3125
Yellow
Case of 500 Each for $36.69
Only null left
 
22-038-504
View Documents
Simport™ Scientific
T31210
Lilac
Case of 500 Each for $32.68
Only null left
 
22-038-502
View Documents
Simport™ Scientific
T31214
Pink
Case of 500 Each for $34.66
Only null left
 
36-101-5340
View Documents
Simport™ Scientific
T3121
White
Case of 500 Each for $36.47
Only null left
 
23-045-226
View Documents
Simport™ Scientific
T3122
Blue
Pack of 500 Each for $32.68
Only null left
 
23-045-227
View Documents
Simport™ Scientific
T3123
Red
Pack of 500 Each for $32.68
Only null left
 
23-045-228
missing translation for 'viewDocuments'
Simport™ Scientific
T3124
Green
Pack of 500 Each for $32.68
Only null left
 
Description

Description

  • Color coded inserts
  • Made of polypropylene
  • For Cryovial tubes
Specifications

Specifications

Polypropylene
Cryovial™ Tubes
Videos
SDS
Documents

Documents