missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Simport™ Scientific CoreDish™ Biopsy Containers

CoreDish™ Biopsy Containers allow for safe and easy transport of specimens from collection to analysis.

Supplier:  Simport™ Scientific M970D5B2

 View more versions of this product

Catalog No. 22-045-670


Explore available promotions

Description

Description

Performing a large number of biopsies means an increase in the number of containers handled, and consequently a technical overload of the transmission network, which occurs without any financial counterpart. A new approach had to be developed in order to increase productivity. The CoreDish is a multi-compartment container in the shape of a dish and half prefilled with 10% Neutral Buffered Formalin, for holding and transporting biopsies. Thanks to this innovative biopsy container, it is no longer necessary to use a multitude of individual containers, thereby reducing risk of confusion.

  • Multi-compartment container is constructed of polystyrene
  • Leakproof screw closure with O-ring ensures total protection of contents
  • Writing area for patient information is provided on the label
  • Each compartment is clearly identified to allow proper placement and visualization of the biopsy being inserted
  • Dimensions: 15 H x 95 mm dia.
General Purpose
  • For up to 5, 8 or 12 biopsies (depending on model)
  • Compartments are numbered from 1 to 5, 1 to 8, or 1 to 12
Breast
  • For breast biopsies
  • Out of five compartments, four are labelled: LUQ, RUQ, LLQ, RLQ

Prostate
  • A multi-compartment container for prostate biopsies
  • Eight compartment; six are labeled: Base, Lateral Base, Medium, Lateral Medium, Apex, Lateral Apex
  • 12 compartment; all are labeled: L Base, R Base, L Lateral Base, R Lateral Base, L Lateral Medium, L Medium, R Medium, R Lateral Medium, L Lateral Apex, L Apex, R Apex, R Lateral Apex
Upper GI Tract
  • For upper GI track biopsies
  • Eight compartments; seven are labeled: Gastric Card, Gastric Body, GE Junction, Gastric ATR, Distal Oesophage, Pylorus, Duodeum
Lower GI Tract
  • For lower GI track biopsies
  • Twelve compartments; ten are labeled: Proximal Traverse Colon, Hepatic Flexure Colon, Distal Transverse Colon, Ascending Colon, Splenic flexure Colon, Cecum, Decending Colon, Terminal Ileum, Rectum, Sigmoid Colon

Specifications

Specifications

Polystyrene
Leakproof Screw Cap with O-Ring
15 mm
Breast
Conforms to OSHA directives.
95 mm
5 Compartments
Videos
SDS
Documents

Documents

Promotions

Promotions