Simport™ Scientific CytoSep™ Funnels for Shandon™ Cytospin 4™ Centrifuges
CytoSep™ Funnels for Shandon™ Cytospin 4™ Centrifuges for use with Thermo Scientific™ Shandon Cytospin 4 Centrifuges.
$568.34 - $2013.03
Catalog Number | Mfr. No. | Type | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity | ||||||
22-045-303
|
Simport™ Scientific
M9641 |
Single Funnel Only |
Case of 500 Each for $568.34
|
|
||||||
22-045-301
|
Simport™ Scientific
M96410FW |
Single Funnel with White Filter and Cap |
Case of 500 Each for $1,095.95
|
|
||||||
22-045-302
|
Simport™ Scientific
M96410FT |
Single Funnel with Brown Filter and Cap |
Case of 500 Each for $1,132.24
|
|
||||||
22-045-308
|
Simport™ Scientific
M9641D |
Double Cytology Funnel Only |
Case of 500 Each for $1,001.87
|
|
||||||
22-045-311
|
Simport™ Scientific
M96510FT |
Single Funnel with Brown Filter and Cap, All Plastic |
Case of 480 Each for $1,643.03
|
|
||||||
22-045-307
|
Simport™ Scientific
M96420FW |
Double Funnel w/ White Filter and Cap |
Case of 500 Each for $1,481.61
|
|
||||||
22-045-312
|
Simport™ Scientific
M9651 |
Single Funnel Only, All Plastic |
Case of 500 Each for $814.79
|
|
||||||
22-045-314
|
Simport™ Scientific
M96520FW |
Double Funnel w/ White Filter and Cap, All Plastic |
Case of 480 Each for $2,013.03
|
|
||||||
22-045-315
|
Simport™ Scientific
M9651D |
Double Cytology Funnel Only, All Plastic |
Case of 500 Each for $844.87
|
|
||||||
M965-10FW
|
Simport™ Scientific
M96510FW |
Single Funnel with White Filter and Cap, All Plastic |
Case of 480 Each for $1,643.03
|
|
||||||
Description
- All plastic disposable sample chambers do not need metal clips
- Available with or without pre-attached filter card
Specifications
Shandon Cytospin 4 |