Simport™ Scientific EasyDip™ Slide Staining Jars
User-friendly approach for staining microscope slides. These EasyDip™ Slide Staining Jars are a part of the EasyDip™ Slide Staining System. (See also the EasyDip Slide Staining Rack.)
Supplier:  Simport™ Scientific M90012G
 View more versions of this product
Catalog No. 22-038-490
Explore available promotions
Description
Description
  • Consists of square staining jar and a vertical slide rack (sold separately, M90512DGY)
  • Vertical slide staining rack will hold up to 12 standard microscope slide
  • Slide writing area is kept out of staining fluid
  • Jars can be loosely joined to each other
  • Made of acetal plastic that resists attacks by most staining agents (except phenol, iodine or ferric chloride)
Specifications
Specifications
Not Autoclavable
80 mL
Acetal
Special stains, frozen sections and special processes
12 standard microscope slides
76 mm
64 mm
3.625 in.
Flat
Green
Slide Staining Jar
6/Cs.
3 in.
2.5 in.
92 mm