missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Simport™ Scientific Polyethylene Flange Plug Caps

These Flange Plug Caps have two flexible flanges to ensure a leakproof seal. They will fit into test tubes and also in round cuvettes and centrifuge tubes.

Supplier:  Simport™ Scientific T40410GDUP

 View more versions of this product

Catalog No. 22-038-139


Explore available promotions

Description

Description

  • Made of polyethylene
  • Easy-on, easy-off design on 12 m and 16 mm OD tubes
  • Protects from aerosols of highly infectious microorganisms
  • Available in 6 colors
Specifications

Specifications

Flange Plug Cap
Green
16 mm Tubes
Polyethylene
1000 Pk.
Videos
SDS
Documents

Documents

Promotions

Promotions