missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mopec Single Faucet Autopsy Scrub Sink

Essential product for any anatomy lab providing a specific place to wash hands before and after an autopsy or any handling of infectious material. Mopec™ Single Faucet Autopsy Scrub Sink has no parting seams which can potentially collect bacteria.

Supplier:  Mopec MPCCH100

Catalog No. 22444042

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

  • Stainless steel scrub sink designed with a 12 in. backsplash
  • Hot and cold foot pedal valve with gooseneck faucet

Warranty

1 Year Warranty

Specifications

Specifications

Autopsy and Embalming Sink
19 in.
48.7 cm
30 in.
58.5 cm
68 cm
76.2 cm
Stainless Steel
11 in.
28 cm
23 in.
26.75 in.
15.37 in.
39 cm
Videos
SDS
Documents

Documents

Promotions

Promotions